Learn More
Sodium phosphate dodecahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Description
Sodium phosphate dodecahydrate is used in photographic developing, clarifying sugar, removing boiler scale, softening water and manufacturing of paper. They are added to many foods as an emulsifier to prevent oil separation. It can be utilized as a leavening agent. Some instances of these foods include the batter coating on breaded fish or chicken, and commercially baked cakes. Adding this item to food increases the shelf life of the food, maintaining the texture and visual aspect of the food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼75°C (decomposition) |
Color | White |
pH | 12 |
Physical Form | Crystalline |
Quantity | 500 g |
Linear Formula | Na3PO4·12H2O |
UN Number | UN3262 |
Merck Index | 14,8662 |
Solubility Information | Soluble in water; Insoluble in alcohol. |
Formula Weight | 380.12 (163.94 Anhydrous) |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S. |
EINECSNumber | 231-509-8 |
RTECSNumber | TC9575000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.