Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic, Anhydrous, Purified (Sodium Dihydrogen Phosphate), Reagents

Catalog No. 01336989
Click to view available options
Quantity:
500 g
3 kg
10 kg

Sodium Phosphate Monobasic, Anhydrous, Purified (Sodium Dihydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym Sodium Dihydrogen Phosphate
IUPAC Name sodium dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Boiling Point 204°C
Color White
Physical Form Solid
Quantity 3 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.04
Chemical Name or Material Sodium Phosphate Monobasic, Anhydrous, Purified (Sodium Dihydrogen Phosphate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.