Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic, Monohydrate, Granular, FCC, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18606714
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

S1397, 10049-21-5, NaH2PO4·H2O

Spectrum™ Chemical Sodium Phosphate Monobasic, Monohydrate, Granular, FCC is an additive used as an antioxidant, sequestrant and a pH regulator with very high buffering capacity widely used in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 98 to 103%
Grade FCC
Loss on Drying 10-15%
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.