Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate, tribasic dodecahydrate, 98%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
25kg
Packaging:
Fibre drum
Plastic bottle
10101-89-0
Na3O412H2O
380.13
MFCD00149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate,sodium phosphate dodecahydrate,sodium phosphate tribasic dodecahydrate,phosphoric acid, trisodium salt, dodecahydrate,unii-b70850qphr,ccris 7322,sodium phosphate, tribasic, dodecahydrate,trisodium dodecahydrate phosphate,phosphoric acid, trisodium salt, dodeahydrate,trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
10101-89-0
Na3O412H2O
380.13
MFCD00149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate,sodium phosphate dodecahydrate,sodium phosphate tribasic dodecahydrate,phosphoric acid, trisodium salt, dodecahydrate,unii-b70850qphr,ccris 7322,sodium phosphate, tribasic, dodecahydrate,trisodium dodecahydrate phosphate,phosphoric acid, trisodium salt, dodeahydrate,trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
Biological buffer
Assay Percent Range 98%
Linear Formula Na3PO4·12H2O
Merck Index 15, 8792
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 380.13
Physical Form Crystalline Powder, Needles or Crystals
Percent Purity 98%
Grade Analysis
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Density 1.6200g/mL
Free Alkali 2.5% max.
Iron (Fe) 50ppm max.
Lead (Pb) 20ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
Specific Gravity 1.62
Sulfate 100ppm max
Total Sulfur 200ppm max.
Color White
Melting Point 75.0°C
Quantity 1kg
Chemical Name or Material Sodium phosphate, tribasic dodecahydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning