Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate, tribasic dodecahydrate, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
10101-89-0
Na3O412H2O
380.13
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate,sodium phosphate dodecahydrate,sodium phosphate tribasic dodecahydrate,phosphoric acid, trisodium salt, dodecahydrate,unii-b70850qphr,ccris 7322,sodium phosphate, tribasic, dodecahydrate,trisodium dodecahydrate phosphate,phosphoric acid, trisodium salt, dodeahydrate,trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
10101-89-0
Na3O412H2O
380.13
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate,sodium phosphate dodecahydrate,sodium phosphate tribasic dodecahydrate,phosphoric acid, trisodium salt, dodecahydrate,unii-b70850qphr,ccris 7322,sodium phosphate, tribasic, dodecahydrate,trisodium dodecahydrate phosphate,phosphoric acid, trisodium salt, dodeahydrate,trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
Linear Formula Na3PO4·12H2O
Merck Index 15,8792
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 380.13
Grade Extra Pure
Density 1.6200g/mL
Insoluble Matter 0.05% max. (in water)
Packaging Plastic bottle
Solubility Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
Specific Gravity 1.62
Melting Point 75.0°C
Quantity 1kg
Chemical Name or Material Sodium phosphate, tribasic dodecahydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning