Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate Tribasic, Dodecahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical

Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Sodium Phosphate Tribasic, Dodecahydrate, Crystal, Reagent, ACS is one of the sodium salts derived from phosphoric acid. Highly hygroscopic and water soluble it comes in the form of a white powder. Used commercially in powered product as an anticaking agent for those qualities. In an aqueous solution it is basic, with a pH value between 8.0 and 11.0. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
CAS Max % | 1 |
Quantity | 12 kg |
Percent Purity | 98 to 102% |
Grade | ACS |
Packaging | Poly Pail |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction