Learn More
Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL
Description
Sodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Physical Form | Liquid |
Solubility | Miscible with water |
Quantity | 500 mL |
Concentration or Composition (by Analyte or Components) | Nitric acid: 5%; Sodium carbonate: 2.3%; Water: 92.7% |
Chemical Name or Material | Sodium, plasma standard solution |
Safety and Handling
Hazard Category | H290-H314-H335 |
Hazard Statement | Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. |
Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
DOTInformation | DOT Class: 8, Packaging Group: III |
EINECSNumber | 231-714-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.