Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Potassium ATPase Alpha 1 Rabbit anti-Human, Mouse, Rat, Clone: 3L6F0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Sodium Potassium ATPase Alpha 1 |
---|---|
Clone | 3L6F0 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Sodium Potassium ATPase Alpha 1 Monoclonal antibody specifically detects Sodium Potassium ATPase Alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Sodium Potassium ATPase Alpha 1 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ATPase, Na+/K+ transporting, alpha 1 polypeptide, EC 3.6.3, EC 3.6.3.9, K-ATPase alpha-1 subunit, K-ATPase catalytic subunit alpha-A protein, MGC3285, MGC51750, Na, Na(+)/K(+) ATPase alpha-1 subunit, Na, K-ATPase, alpha-A catalytic polypeptide, Na+, K+ ATPase alpha subunit, Na+/K+ ATPase 1, sodium pump 1, Sodium pump subunit alpha-1, sodium/potassium-transporting ATPase subunit alpha-1, sodium-potassium-ATPase, alpha 1 polypeptide | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Sodium Potassium ATPase Alpha 1 (P05023). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3L6F0 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
476 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title