Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Potassium ATPase Alpha 3 Antibody (XVIF9-G10), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300540PECY55
Description
Sodium Potassium ATPase Alpha 3 Monoclonal specifically detects Sodium Potassium ATPase Alpha 3 in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Flow Cytometry.Specifications
Sodium Potassium ATPase Alpha 3 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
ATP1A3 | |
Canine cardiac microsomes. | |
0.1 mL | |
478.0 | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Rabbit, Sheep | |
IgG1 |
Flow Cytometry | |
XVIF9-G10 | |
Flow Cytometry | |
alpha 3 polypeptide, ATPase, Na+/K+ transporting, alpha 3 polypeptide, dystonia 12, DYT12, EC 3.6.3, MGC13276, RDP, Sodium pump subunit alpha-3, sodium/potassium-transporting ATPase alpha-3 chain, sodium/potassium-transporting ATPase subunit alpha-3 | |
Mouse | |
Protein A purified | |
Primary | |
Detects sodium/ potassium ATPase. This is specific for the alpha-3 subunit. | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction