Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Potassium Tartrate, 25% (w/v), Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Color | Colorless |
CAS Min % | 78.0, 19.61 |
CAS Max % | 82.0, 20.41 |
Percent Purity | 25% |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
pH | 7 to 8 |
Quantity | 1 L |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction