Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium propionate, 99%

Catalog No. AAA174400E
Click to view available options
Quantity:
500 g
2500 g

CAS: 137-40-6 | C3H5NaO2 | 96.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium propionate is used as a food preservative, as a mold inhibitor in bakery products and in insect cell culture. It is also used in pharmaceuticals, cosmetics, agriculture and animal feed as preservative. It acts as a food additive. Further, it inhibits the growth of lactic acid bacteria.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-40-6
Molecular Formula C3H5NaO2
Molecular Weight (g/mol) 96.061
MDL Number MFCD00002759
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
Synonym sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
PubChem CID 2723816
IUPAC Name sodium;propanoate
SMILES CCC(=O)[O-].[Na+]

Specifications

Melting Point 285°C to 291°C
Assay Percent Range 99%
Linear Formula CH3CH2CO2Na
Quantity 2500 g
Sensitivity Hygroscopic
Merck Index 14,8669
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 96.06
Percent Purity 99%
Chemical Name or Material Sodium propionate
Hazard Category Warning
Hazard Statement GHS H Statement
H312
Harmful in contact with skin.
Precautionary Statement GHS P Statement
P280-P312-P363-P322-P302+P352-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 205-290-4
RTECSNumber UF7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.