Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pyrophosphate decahydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
10kg
13472-36-1
Na4O7P2·10H2O
446.05
MFCD00149200
VZWGHDYJGOMEKT-UHFFFAOYSA-J
sodium pyrophosphate decahydrate,unii-iy3dkb96qw,tetrasodium pyrophosphate decahydrate,diphosphoric acid, tetrasodium salt, decahydrate,iy3dkb96qw,pyrophosphate sodium salt decahydrate,pyrophosphate inorganic,colostrum,sodium phosphate pyro,sodium pyrophosphate na4p2o7 decahydrate
3084150
tetrasodium;phosphonato phosphate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
13472-36-1
Na4O7P2·10H2O
446.05
MFCD00149200
VZWGHDYJGOMEKT-UHFFFAOYSA-J
sodium pyrophosphate decahydrate,unii-iy3dkb96qw,tetrasodium pyrophosphate decahydrate,diphosphoric acid, tetrasodium salt, decahydrate,iy3dkb96qw,pyrophosphate sodium salt decahydrate,pyrophosphate inorganic,colostrum,sodium phosphate pyro,sodium pyrophosphate na4p2o7 decahydrate
3084150
tetrasodium;phosphonato phosphate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
Biological buffer
Additional Information ortho-phosphate (as PO4): < 0.1%
Assay Percent Range 99+%
Linear Formula Na4P2O7·10H2O
Merck Index 10,9063
Solubility Information Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia,insoluble in alcohol
Formula Weight 446.05
Physical Form Crystals
Percent Purity 99+%
Grade Analysis
Chloride 10ppm max
Chlorine (Cl) 10ppm max.
Density 1.82
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 5ppm max.
Packaging Plastic bottle
Potassium (K) 50ppm max.
Total Nitrogen Compounds 0.001% max. (as N)
Boiling Point 93.8°C
Color White
Melting Point 80.0°C
Quantity 500g
Chemical Name or Material Sodium pyrophosphate decahydrate, p.a.
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning