Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Pyrophosphate Decahydrate, ACS Reagent, Reagents
Click to view available options
Quantity:
500 g
2.5 kg
10 lb.
10 kg
Specifications
Specifications
Chemical Name or Material | Sodium Pyrophosphate Decahydrate, ACS Reagent |
Boiling Point | 93°C |
Color | Colorless/White |
CAS | 13472-36-1 |
Physical Form | Solid |
Quantity | 10 kg |
Molecular Formula | Na4P2O7, 10H2O |
Synonym | Tetrasodium Diphosphate Decahydrate |
Molecular Weight (g/mol) | 446.0552 |
Grade | ACS |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction