Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium salicylate, 99%

Catalog No. aaa1705630
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1705630 250 g
AAA170560B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1705630 Supplier Thermo Scientific Chemicals Supplier No. A1705630
Only null left
Add to Cart

CAS: 54-21-7 | C7H5NaO3 | 160.104 g/mol

Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

Solubility
Soluble in water, 1,4-dioxane, ethanol and glycerol.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acidsmineral acids, metallic salts and iodine.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
MDL Number MFCD00002440
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
PubChem CID 16760658
ChEBI CHEBI:9180
IUPAC Name sodium;2-hydroxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]

Specifications

Melting Point >300°C
Odor Odorless
Quantity 250 g
Beilstein 3732792
Sensitivity Light sensitive
Merck Index 14,8332
Solubility Information Soluble in water,1,4-dioxane,ethanol and glycerol.
Formula Weight 160.11
Percent Purity 99%
Chemical Name or Material Sodium salicylate
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 200-198-0
RTECSNumber VO5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.