Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Salicylate, Crystal, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606777 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail

Catalog No. 18-606-777

Supplier: Spectrum Chemical Manufacturing Corporation SO20512KG

Only null left
Add to Cart
Add to Cart

SO205, 54-21-7, C7H5O3Na

Spectrum™ Chemical Sodium Salicylate, Crystal, USP is a non-steroidal anti-inflammatory drug used in medicine as an antipyretic and analgesic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
IUPAC Name sodium 2-hydroxybenzoate
SMILES [Na+].OC1=CC=CC=C1C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.