Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Salicylate Nitroprusside Solution, Reagents

Catalog No. 01333999
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-999 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-999 Supplier Reagents Holdings Llc Supplier No. CS0072004F
Only null left
Add to Cart
Add to Cart

Sodium Salicylate Nitroprusside Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Salicylate Nitroprusside Solution
Boiling Point 100°C
Color Tan
CAS 13755-38-9, 54-21-7, 7732-18-5
Physical Form Liquid
Quantity 4 L
Packaging Cubitainer
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.