Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium selenide, 99.8% (metals basis)

Catalog No. aa3618703
Click to view available options
Quantity:
1 g
5 g

CAS: 1313-85-5 | Na2Se | 124.951 g/mol

Sodium selenide is used in paper and pulp industry, for kraft process followed by photographic industry to prevent developer solutions. It finds application as a bleaching agent in textile industry, sulfur dyes and detergents. It is also used to impart color to glass, glazes and enamels. Further, it acts as a decolorizer of glass due to neutralization of greenish tint produced by iron (ferrous) compounds. It plays an important role to prepare organoselenium compounds by reacting with alkyl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-85-5
Molecular Formula Na2Se
Molecular Weight (g/mol) 124.951
MDL Number MFCD00014240
InChI Key VPQBLCVGUWPDHV-UHFFFAOYSA-N
Synonym sodium selenide, disodium selenide, disodium monoselenide, unii-1fek1k7wce, disodium selandiide, 1fek1k7wce, sodiumselenide, disodium selenium 2-, parent
PubChem CID 73973
ChEBI CHEBI:77773
IUPAC Name disodium;selenium(2-)
SMILES [Na+].[Na+].[Se-2]

Specifications

Melting Point >875°C
Color Gray to Pink
Quantity 1 g
Assay Percent Range 99.8% (metals basis)
UN Number UN3283
Merck Index 14,8672
Solubility Information Soluble in ethanol and liquid ammonia. Slightly soluble tetrahydrofuran.
Formula Weight 124.94
Percent Purity 99.8%
Grade ACS
Sensitivity Air and moisture sensitive; Hygroscopic
Density 2.625 g/mL
Assay (metals basis)
Chemical Name or Material Sodium selenide
Show More Show Less
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H300-H331-H373
Fatal if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
EINECSNumber 215-212-0
RTECSNumber WE0350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.