Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium selenite, 44-46% Se, anhydrous, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
10102-18-8
Na2O3Se
172.94
MFCD00003489
BVTBRVFYZUCAKH-UHFFFAOYSA-L
sodium selenite,disodium selenite,natriumselenit,selenious acid, disodium salt,natriumselenit german,disodium selenium trioxide,unii-hiw548rq3w,selenious acid disodium salt,ccris 1260,hsdb 768
24934
CHEBI:48843
disodium;selenite
[O-][Se](=O)[O-].[Na+].[Na+]
10102-18-8
Na2O3Se
172.94
MFCD00003489
BVTBRVFYZUCAKH-UHFFFAOYSA-L
sodium selenite,disodium selenite,natriumselenit,selenious acid, disodium salt,natriumselenit german,disodium selenium trioxide,unii-hiw548rq3w,selenious acid disodium salt,ccris 1260,hsdb 768
24934
CHEBI:48843
disodium;selenite
[O-][Se](=O)[O-].[Na+].[Na+]
Linear Formula Na2SeO3
Merck Index 15, 8802
Solubility Information Solubility in water: 950g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 172.94
Physical Form Powder
Percent Purity 44-46% Se
Iron (Fe) 300ppm max.
Lead (Pb) 500ppm max.
Loss on Drying 0.5% max.
Packaging Glass bottle
Color White to Beige
Melting Point 350°C
Quantity 100g
Chemical Name or Material Sodium selenite, Anhydrous
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Contact with acids liberates toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
Do not breathe

GHS Signal Word: Danger

EINECSNumber : 233-267-9