Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Selenite, 99.8+%, Powder, Spectrum™ Chemical
SDP

Catalog No. 18613848
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Lined Fiber Drum
Poly Pail

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
IUPAC Name disodium selenite
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

CAS Max % 1
Quantity 25 kg
Percent Purity ≥99.8%
Grade Reagent
Packaging Lined Fiber Drum

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.