Learn More
Sodium Silicate, Technical Grade, 40 to 42°Baumé, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 4 L |
| Molecular Weight (g/mol) | 212.14 g/mol |
| Grade | Technical |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium silicate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.