Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stannate, Granular, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18606789 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-789 500 g Amber Glass Bottle
18-606-788 125 g Amber Glass Bottle
18-606-790 2.5 kg Amber Glass Bottle
18-606-791 12 kg Poly Pail
18-606-792 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-789 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1445500GM
Add to Cart
Add to Cart

S1445, 12027-70-2, Na2SnO3·3H2O

Spectrum™ Chemical Sodium Stannate, Granular, Reagent is a colorless salt that occurs when tin is dissolved in sodium hydroxide. It is an inorganic compound that is used as a hydrogen peroxide stabilizer. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 12027-70-2
Molecular Formula H6Na2O6Sn
Molecular Weight (g/mol) 266.73
InChI Key PMPBLIIMRRPPEO-UHFFFAOYSA-H
IUPAC Name tin(4+) disodium hexahydroxide
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4]

Specifications

CAS Max % 1
Quantity 500 g
Grade Reagent
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.