Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stannate (Sodium Hydroxystannate), Reagents

Catalog No. 01336964
Click to view available options
:
Quantity: 10 kg

Sodium Stannate (Sodium Hydroxystannate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 12027-70-2
Molecular Formula H6Na2O6Sn
Molecular Weight (g/mol) 266.73
InChI Key PMPBLIIMRRPPEO-UHFFFAOYSA-H
IUPAC Name tin(4+) disodium hexahydroxide
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4]

Specifications

Melting Point >140°C
Color White to Tan
Physical Form Solid
Quantity 10 kg
Grade Reagent
Packaging Poly Pail
Specific Gravity 4.68
Chemical Name or Material Sodium Stannate (Sodium Hydroxystannate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.