Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Starch Glycolate, pH 5.5-7.5, Type A, NF, Spectrum™ Chemical
SDP

Catalog No. 18606796 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-796 50 kg Fiber Drum
18-606-794 2.5 kg Poly Pail
18-606-795 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-796 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S196250KGBL
Add to Cart
Add to Cart
Request a Quote

S1962, 9063-38-1

Spectrum™ Chemical Sodium Starch Glycolate, pH 5.5-7.5, Type A, NF is used as a pharmaceutical grade dissolution excipient for tablets and capsules. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9063-38-1

Specifications

CAS Max % 1
Loss on Drying 0.1
Packaging Fiber Drum
Quantity 50 kg
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.