Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stearyl Fumarate, NF, 99-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18606806 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
25 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-806 500 g Poly Pail
18-606-807 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-806 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1448500GM
Add to Cart
Add to Cart

S1448, 4070-80-8, C22H39NaO4

Spectrum™ Chemical Sodium Stearyl Fumarate, NF is a water-soluble tablet lubicant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4070-80-8
Molecular Formula C22H39NaO4
Molecular Weight (g/mol) 390.54
InChI Key STFSJTPVIIDAQX-LTRPLHCISA-M
IUPAC Name sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate
SMILES [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O

Specifications

CAS Max % 1
Melting Point > 200°C
Packaging Poly Pail
Quantity 500 g
Saponification Value 142.2-146.0
Percent Purity 99 to 101.5%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sodium Stearyl Fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.