Learn More
Description
- Sodium succinate dibasic can be used:
- As a liquid phase in the study of basic properties of α-tricalcuim phosphate-based cement
- To prepare succinato-bridged copper(ii) complexes exhibiting magnetic properties
Specifications
Specifications
| CAS | 150-90-3 |
| Molecular Formula | C4H4Na2O4 |
| Linear Formula | NaOOCCH2CH2COONa |
| MDL Number | MFCD00002790 |
| Quantity | 100 g |
| Synonym | Succinic acid disodium salt |
| Molecular Weight (g/mol) | 162.05 |
| Percent Purity | ≥98.0% (NT) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.