Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium succinate hexahydrate, 99%

Catalog No. aa4198330
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 6106-21-4 | C4H16Na2O10 | 270.142 g/mol

Sodium succinate hexahydrate is used as a tricarboxylic acid cycle (Krebs cycle) supplement in cell culture applications. It is also used as a flavoring agent for food and beverages. It serves as an intermediate for dyes, perfumes, lacquers, photographic chemicals, alkyd resins, plasticizer, metal treatment chemical, vehicle water cooling systems and coatings. Further, it is used to study x-ray crystallography, protein structural analysis, proteomics and optimization reagents. It is utilized to determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
MDL Number MFCD00149117
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 120°C to 6H2O
Linear Formula NaO2CCH2CH2CO2Na·6H2O
Quantity 250 g
Beilstein 3921044
Sensitivity Hygroscopic
Merck Index 14,8679
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 270.14 (162.05 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Sodium Succinate Hexahydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-778-7
RTECSNumber WM7751000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.