Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Succinate, Hexahydrate, Reagent, 97-102%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Sodium Succinate, Hexahydrate, Reagent , also known as succinic acid, is a diprotic and dicarboxylic acid. It is involved in an energy-yielding process known as the citric acid cycle. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
CAS Max % | 1 |
Quantity | 12 kg |
Percent Purity | 97 to 102% |
Grade | Reagent |
Packaging | Fiber Drum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction