Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, 98.0%, MilliporeSigma™

Catalog No. 50947728 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Plastic Bottle
Plastic Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50-947-728 12 kg Plastic Drum
50-947-272 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-947-728 Supplier MilliporeSigma Supplier No. SX07635

Chemical Identifiers

CAS 7757-82-6
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 888°C
pH 5.2 to 8.0 (50 g/L, H2O, 20°C)
Quantity 12 kg
Assay Percent Range 98.0% min
Molecular Formula Na2O4S
Solubility Information 200 g/L
Packaging Plastic Drum
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.