Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium dichromate, 99.5%, for analysis
Click to view available options
Quantity:
50 g
500 g
2 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxidation reagent
Specifications
Specifications
| Boiling Point | 500.0°C |
| Melting Point | 398.0°C |
| Color | Orange |
| Physical Form | Fine Crystals or Crystalline Powder |
| Quantity | 2 kg |
| Assay Percent Range | 99.5% min. (Iodometry) |
| Linear Formula | K2Cr2O7 |
| Fieser | 08,410; 09,383; 10,324; 12,405 |
| Merck Index | 15, 7748 |
| Solubility Information | Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Fatal if inhaled. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. May cause allergy or asthma symptoms or breathing difficu |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major |
| EINECSNumber | 231-906-6 |
| RTECSNumber | HX7680000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction