Learn More
Specifications
Specifications
| pH | 5.2 to 9.2 (5% solution at 25°C) |
| Physical Form | Granules |
| Quantity | 12 kg |
| Assay Percent Range | min. 99.0% |
| Formula Weight | 142.04 |
| Grade | For ACS analysis |
| Loss on Ignition | max. 0.5% |
| Packaging | STAKMOR |
| Appearance | White crystalline granules |
| Chemical Name or Material | Sodium Sulfate, Anhydrous, 10-60 Mesh |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.