Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, ACS Reagent, Reagents

Catalog No. 01337681
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
50 lb.
45 kg
Packaging:
Fiber Drum
Glass Bottle
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
01-337-681 3 kg Poly Bottle
01-337-682 10 kg Poly Pail
01-337-680 3 kg Glass Bottle
01-337-679 45 kg Fiber Drum
01-337-678 500 g Glass Bottle
01-337-677 50 lb. Fiber Drum
6 options

Catalog No. 01-337-681

Supplier: Reagents Holdings Llc C13255003A5

Only null left
Add to Cart
Add to Cart

Sodium Sulfate, Anhydrous, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym Sulfuric Acid disodium salt
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >890°C
Melting Point 884°C to 888°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.7
Chemical Name or Material Sodium Sulfate, Anhydrous, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.