Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, Granular, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18606827 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-827 2.5 kg Amber Glass Bottle
18-606-825 500 g Amber Glass Bottle
18-606-826 6 x 500 g Amber Glass Bottle
18-606-828 4 x 2.5 kg Amber Glass Bottle
18-606-829 12 kg Poly Pail
18-606-830 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-827 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S14502.5KGGL
Add to Cart
Add to Cart

S1450, 7757-82-6, Na2SO4

Spectrum™ Chemical Sodium Sulfate, Anhydrous, Granular, Reagent, ACS is the sodium salt of sulfuric acid. It is a white, crystalline solid, and is used on a small scale in laboratories as an inert drying agent. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

pH 5.2 to 9.2
CAS Max % 1
Quantity 2.5 kg
Percent Purity 99%
Grade ACS
Loss on Ignition 0.005
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.