Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Sulfate, Anhydrous, Granular, Reagent, ACS, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Sodium Sulfate, Anhydrous, Granular, Reagent, ACS is the sodium salt of sulfuric acid. It is a white, crystalline solid, and is used on a small scale in laboratories as an inert drying agent. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
pH | 5.2 to 9.2 |
CAS Max % | 1 |
Quantity | 12 kg |
Percent Purity | 99% |
Grade | ACS |
Loss on Ignition | 0.005 |
Packaging | Poly Pail |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction