Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Anhydrous, min.99%, 12-60 Mesh, Granular, Ultra Resi-Analyzed, ACS Reagent, J.T. Baker™

Catalog No. 14650207 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45.5 kg
Packaging:
Glass Bottle
Lined Fiber Drum
Wide Mouth Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
14-650-207 45.5 kg Lined Fiber Drum
02-004-233 500 g Wide Mouth Amber Glass Bottle
14-650-356 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-650-207 Supplier Avantor Supplier No. 337509
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

pH 5.2 to 9.2
Physical Form Granules
Quantity 45.5 kg
IUPAC Name disodium;sulfate
Formula Weight 142.04
Grade Meets ACS reagent chemical requirements
Loss on Ignition max. 0.5%
Packaging Lined Fiber Drum
Specific Gravity 1L = 2.68kg
Chemical Name or Material Sodium Sulfate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.