Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Solstice

Catalog No. 6001996 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-96 2.5 kg
60-019-95 1 kg
60-044-811 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-019-96 Supplier Solstice Supplier No. 134642.5KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous
Quantity 2.5 kg
Linear Formula Na2SO4
UN Number NONH for all modes of transport
IUPAC Name disodium;sulfate
Formula Weight 142.04g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium Sulfate, Meets analytical specification of Ph. Eur.Bp, USP
EINECSNumber 231-820-9
RTECSNumber WE1650000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.