Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, 5% w/v, Reagents

Catalog No. 01333914
Change view
Click to view available options
:
Quantity: 500 mL
Catalog No.
01-333-914 Quantity: 500 mL
1 options

Catalog No. 01-333-914

Supplier: Reagents Holdings Llc CS023000500C

Sodium Sulfide, 5% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1313-84-4
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 500 mL
Percent Purity 5%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0463
Chemical Name or Material Sodium Sulfide, 5% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.