Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals

Catalog No. p-42394
Click to view available options
Quantity:
10 g
250 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-82-2
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00003498
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
PubChem CID 237873
SMILES [Na+].[Na+].[S--]

Specifications

Color Beige-Yellow
Physical Form Lump/Powder
Quantity 10 g
Assay Percent Range 90% min. (Iodimetry)
Linear Formula Na2S
Merck Index 15,881
Solubility Information Solubility in water: 186g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether
IUPAC Name disodium;sulfanide
Formula Weight 78.04
Percent Purity ≥90%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium sulfide, 90%, balance water
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosive
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 215-211-5
RTECSNumber WE1905000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.