Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals

Catalog No. p-3735169
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Fibre drum
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
PubChem CID 45051681
SMILES [Na+].[Na+].[S--]

Specifications

Color Yellow
Physical Form Flakes and Chunks
Quantity 1 kg
Assay Percent Range 60 to 64% (Na2S) (Iodimetry)
Linear Formula Na2S·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether
IUPAC Name disodium sulfanediide
Formula Weight 78.04
Percent Purity 60 to 64%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium sulfide hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosiv
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.