Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Sulfide Nonahydrate
SCI_ED

Catalog No. p-4416478 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
Synonym Sodium Monosulfide Nonahydrate
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

Color Yellow
Quantity 100 g
Assay Percent Range >95%
Linear Formula Na2S·9H2O
Formula Weight 240.18
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Sodium Sulfide, Nonahydrate
ShelfLife 1 year
DOTInformation DOT Class 8, II, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.