Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfite, 98+%, ACS reagent, anhydrous

Catalog No. p-3593525
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC424432500

Supplier: Thermo Scientific Chemicals 424432500

Only null left
Add to Cart

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 250 g
Assay Percent Range 98% min. (Iodimetry)
Linear Formula Na2SO3
Fieser 02,387
Merck Index 15, 8811
Solubility Information Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
Formula Weight 126.04
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.63
Decomposition Information >500°C
Density 2.6300g/mL
Chemical Name or Material Sodium sulfite
Show More Show Less
Hazard Statement GHS H Statement:
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-821-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.