Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, ACS Reagent, Reagents

Catalog No. 01337670
Click to view available options
Quantity:
500 g
2.5 kg
10 kg

Sodium Sulfite, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula H14Na2O10S
Molecular Weight (g/mol) 252.14
InChI Key WMJRPJZQQSSDBU-UHFFFAOYSA-L
Synonym Sulfurous Acid disodium salt
IUPAC Name disodium heptahydrate sulfite
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O

Specifications

Melting Point >600°C
Color White
Physical Form Solid
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 2.633
Chemical Name or Material Sodium Sulfite, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.