Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Tartrate, 25% w/v, pH 7, Reagents
Sodium Tartrate, 25% w/v, pH 7, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CS01320020F
Quantity | 20 L |
---|
Chemical Identifiers
7732-18-5, 6106-24-7 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
Colorless | |
102°C | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
25% | |
Cubitainer | |
7 | |
Non-sterile | |
Sodium Tartrate, 25% w/v, pH 7 |
7732-18-5, 6106-24-7 | |
H2O | |
O | |
18.02 | |
Reagent | |
1.1352 | |
20 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction