Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium taurocholate hydrate, 96%

Catalog No. p-3173441
Click to view available options
Quantity:
1 g
5 g
25 g

Sodium taurocholate hydrate (CAS # 145-42-6) is a bile acid and detergent used in molecular biology for protein solubilization. | CAS: 345909-26-4 | C26H44NNaO7S | 537.688 g/mol

Sodium taurocholate is a bile acid having detergent capabilities; natural surfactant. It is used for solubilization of protein. In the preparation of digestive enzymes, it is used as an emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium taurocholate hydrate is a bile acid conjugate of taurine and cholic acid that acts as a detergent
  • Sodium taurocholate solubilizes fats within the digestive system
  • It is the main component of bile acid in carnivorous animals; metabolites are also common in human bile

Application

  • Sodium taurocholate hydrate can be used as an emulsifier for protein isolation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-26-4
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.688
MDL Number MFCD00150819
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
Synonym sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
PubChem CID 131632374
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Assay Percent Range 0.97
Quantity 1 g
Solubility Information Soluble in Water (50mg/mL- Clear Solution)
Formula Weight 537.73 (Anhydrous)
Chemical Name or Material Sodium taurocholate hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-653-7
RTECSNumber KI8019250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.