Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tert-butoxide, 99%, pure

Catalog No. p-3737358
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC252550050 5 g Glass bottle
AC252551000 100 g Glass bottle
AC252555000 500 g Glass bottle
AC252550025 2.5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC252550050 Supplier Thermo Scientific Chemicals Supplier No. 252550050
Only null left
Add to Cart

CAS: 865-48-5 | C4H9NaO | 96.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-48-5
Molecular Formula C4H9NaO
Molecular Weight (g/mol) 96.1
MDL Number MFCD00040575
InChI Key MFRIHAYPQRLWNB-UHFFFAOYSA-N
Synonym sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
PubChem CID 23676156
IUPAC Name sodium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[Na+]

Specifications

Melting Point 180.0°C
Flash Point 12°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NaOC(CH3)3
Quantity 5 g
Fieser 16,55
Solubility Information Solubility in water: reacts.
Formula Weight 96.1
Percent Purity 99%
Grade Pure
Chemical Name or Material Sodium tert-butoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Self-heating; may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.
Flammable solid.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
EINECSNumber 212-741-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.