Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloroaluminate

Catalog No. p-4407944
Click to view available options
Quantity:
25 g
100 g

CAS: 7784-16-9 | AlCl4Na | 191.771 g/mol

Sodium tetrachloroaluminate is used as Friedel-Crafts acylating agent , dehydrating and dehydrogenating agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-16-9
Molecular Formula AlCl4Na
Molecular Weight (g/mol) 191.771
MDL Number MFCD00050361
InChI Key CMLRNXGMTZKKSR-UHFFFAOYSA-J
Synonym sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis
PubChem CID 16699350
IUPAC Name sodium;tetrachloroalumanuide
SMILES [Na+].[Al-](Cl)(Cl)(Cl)Cl

Specifications

Melting Point 185°C
Color Yellow
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in water
Formula Weight 191.78
Sensitivity Moisture sensitive
Appearance Pale yellow powder
Density 2.01 g/cm3
Chemical Name or Material Sodium tetrachloroaluminate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8, Packing Group: III
EINECSNumber 232-050-6
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.