Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%

Catalog No. p-4412865
Click to view available options
Quantity:
1 g
5 g

CAS: 13874-02-7 | AuCl4H4NaO2 | 397.79 g/mol

It is used in chemo-therapeutics. It is employed as a catalyst in the removal of tert-butyl(dimethyl)silyl (TBS) protecting groups. It is selective in the presence of aromatic TBS ethers, aliphatic triisopropylsilyl ethers, aliphatic tert-butyl(diphenyl)silyl ethers, or sterically hindered aliphatic TBS ethers. With excess catalyst, TBS ethers can also be transformed into 4-methoxybenzyl ethers or methyl ethers in one pot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13874-02-7
Molecular Formula AuCl4H4NaO2
Molecular Weight (g/mol) 397.79
MDL Number MFCD00149162
InChI Key OEERDTKOVZFOHF-UHFFFAOYSA-J
Synonym sodium tetrachloroaurate iii dihydrate, sodium;tetrachlorogold 1-;dihydrate, sodiumtetrachloroaurate iii dihydrate, sodium dihydrate tetrachloroaurate
PubChem CID 18949434
SMILES O.O.[Na+].Cl[Au-](Cl)(Cl)Cl

Specifications

Melting Point 100°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water,alcohol and ether.
IUPAC Name sodium tetrachlorogolduide dihydrate
Formula Weight 397.80 (361.77 Anhydrous)
Density 0.8 g/mL
Chemical Name or Material Sodium tetrachloroaurate(III) dihydrate
Hazard Category H290-H302-H314-H335
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 239-241-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.