Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloropalladate(II), approx. 36% Pd, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13820-53-6
Cl4Na2Pd
294.21
MFCD00003487
ZIXLZKBNIAXVBE-UHFFFAOYSA-J
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
11000870
disodium;tetrachloropalladium(2-)
[Na+].[Na+].Cl[Pd-2](Cl)(Cl)Cl
13820-53-6
Cl4Na2Pd
294.21
MFCD00003487
ZIXLZKBNIAXVBE-UHFFFAOYSA-J
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
11000870
disodium;tetrachloropalladium(2-)
[Na+].[Na+].Cl[Pd-2](Cl)(Cl)Cl
Assay Percent Range 36%
Linear Formula Na2PdCl4
Fieser 05,626; 06,45; 09,207; 16,309; 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Solubility Information Solubility in water: soluble
Formula Weight 294.21
Physical Form Powder and Granules Crystals
Percent Purity approx. 36% Pd
Calcium (Ca) 10ppm max.
Copper (Cu) 10ppm max.
Gold (Au) 20ppm max.
Iridium (Ir) 20ppm max.
Iron (Fe) 50ppm max.
Magnesium (Mg) 10ppm max.
Packaging Glass bottle
Platinum (Pt) 20ppm max.
Rhodium (Rh) 20ppm max.
Ruthenium (Ru) 20ppm max.
Silicon (Si) 30ppm max.
Silver (Ag) 20ppm max.
Solubility Solubility in water: soluble.
Total Trace Metal Impurities 300ppm (Pt+ Ru+ Rh+ Ir) max
Color Brown to Orange or Brown-Green
Quantity 1g
Chemical Name or Material Sodium tetrachloropalladate(II), ca. 36.4% Pd
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 237-502-6

RTECSNumber : RT3480000

TSCA : TSCA