Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloropalladate(II), approx. 36% Pd

Catalog No. p-42437
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Molecular Weight (g/mol) 294.20
MDL Number MFCD00003487
InChI Key ABKQFSYGIHQQLS-UHFFFAOYSA-J
Synonym sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
PubChem CID 11000870
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

Specifications

Color Brown to Orange or Brown-Green
Physical Form Powder and Granules, Crystals
Quantity 1 g
Assay Percent Range 36%
Linear Formula Na2PdCl4
Fieser 05,626; 06,45; 09,207; 16,309; 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Solubility Information Solubility in water: soluble
IUPAC Name disodium;tetrachloropalladium(2-)
Formula Weight 294.21
Percent Purity ∼36% Pd
Packaging Glass bottle
Chemical Name or Material Sodium tetrachloropalladate(II), ca. 36.4% Pd
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 237-502-6
RTECSNumber RT3480000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.