Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min

Catalog No. p-7059094
Click to view available options
Quantity:
1 g

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol

Sodium tetrachloropalladate is used in organ metallic chemistry for the preparation of phosphine complexes of palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Molecular Weight (g/mol) 294.20
MDL Number MFCD00003487
InChI Key ABKQFSYGIHQQLS-UHFFFAOYSA-J
Synonym sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
PubChem CID 11000870
IUPAC Name disodium;tetrachloropalladium(2-)
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

Specifications

Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Linear Formula Na2PdCl4·xH2O (x≈3)
Solubility Information Soluble in water and ethyl alcohol.
Formula Weight 294.20 (Anhydrous)
Percent Purity 99.999%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pd 30% min
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Sodium tetrachloropalladate (II) hydrate
Show More Show Less
Hazard Category H290-H302-H319
Hazard Statement GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
Precautionary Statement P234-P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 237-502-6
RTECSNumber RT3480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.