Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraethylborate, 98%

Catalog No. AA8913706
Click to view available options
Quantity:
1 g
5 g

CAS: 15523-24-7 | C8H20BNa | 150.048 g/mol

Sodium tetraethylborate is used as a derivatizing agent for the detection of organotin, lead and mercury compounds by using gas chromatography coupled with various sensitive and selective detection methods, such as atomic absorption spectrometry (AAS), atomic emission spectrometry (AES), pulsed-flame photometric detection (PFPD), and mass spectrometry (MS). It is also used as an alkylating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15523-24-7
Molecular Formula C8H20BNa
Molecular Weight (g/mol) 150.048
MDL Number MFCD00061547
InChI Key SZSBMTRYJRHYNI-UHFFFAOYSA-N
Synonym sodium tetraethylborate, sodium tetraethylboranuide, borate 1-,tetraethyl-, sodium 1:1, sodium tetraethylborate 1-, et4bna, sodiotetraethylboron v, borate 1-, tetraethyl-, sodium, sodium tetraethylborate 1g
PubChem CID 23681030
IUPAC Name sodium;tetraethylboranuide
SMILES [B-](CC)(CC)(CC)CC.[Na+]

Specifications

Melting Point (decomposition)
Color White
Assay Percent Range 98%
Linear Formula NaB(C2H5)4
Quantity 5 g
UN Number UN3393
Sensitivity Air and moisture sensitive
Solubility Information Dissolves in water with decomposition
Formula Weight 150.05
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Sodium tetraethylborate
Show More Show Less
Hazard Category H250-H261
Hazard Statement GHS H Statement
H250-H260-H252
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC, WATER-REACTIVE
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.